Would you like to log in again?
Chicago Latrobe® Screw Machine Length Drills, Cutting Tool Supply
Supplier: Cutting Tool Supply
48702 48716 48505 48522 48730 48531 48508 48734 48701 48740 48512 48731 48524 48532 48521 48707 48757 48708 48519 48704 48705 48528 48760 48710 48715 48527 48529 48724 48743 48721 48530 48751 48728 48717 48526
76244-464EA 29.08 USD InStock
76244-464 76244-586 76244-544 76244-566 76244-588 76244-526 76244-548 76244-460 76244-582 76244-462 76244-584 76254-278 76244-538 76254-276 76254-294 76244-574 76244-530 76244-454 76244-532 76244-554 76254-280 76244-456 76244-534 76244-556 76244-458 76244-558 76244-536 76244-590 76244-592 76244-570 76244-450 76244-452 76254-304 76244-528 76254-288
Chicago Latrobe® Screw Machine Length Drills, Cutting Tool Supply
Tool Sets
These general-purpose screw machine length drills are designed for drilling a wide variety of materials on different range of operating conditions.
  • Made of premium high-speed steel
  • 118°-point style
  • Extra rigid for portable drilling
  • Available in fractional, letter and wire gage size
  • Style 157 is either of bright finish or black oxide coated
The shorter flute and overall length provide superior rigidity to minimize deflection when using heavy fee rates. Bright finish with polished flutes enhances chip ejection especially for aluminum and other non-ferrous materials.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty