Would you like to log in again?
Conductive Caps, CEC Series, Caplugs
Supplier: Caplugs
CEC-10 CEC-24 CEC-16 CEC-14 CEC-08 CEC-26 CEC-22 CEC-04 CEC-05 CEC-12
76246-392PK 319.48 USD InStock
76246-392 76246-394 76277-282 76244-996 76246-390 76246-400 76244-282 76244-274 76246-396 76246-398
Conductive Caps, CEC Series, Caplugs
Closures Safety Caps and Safety Waste Caps
CEC series conductive caps are for all common sizes of threaded electrical and electronic connectors. Also suitable for protecting non-threaded bayonet type connectors. In addition to dissipating electrostatic charges, they provide an electrical shield over the end of the connector.
  • Conductive polyethylene copolymer
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty