Would you like to log in again?
Drill Monster® Jobbers Length Twist Drills, Cutting Tool Supply
Supplier: Cutting Tool Supply
450-101875 450-202570 450-301285 450-300550 450-301890 450-300670A 450-101250 450-301040 450-101562 450-301610 450-203970
76249-246EA 46.17 USD InStock
76249-246 76250-102 76249-244 76251-334 76250-684 76249-250 76250-100 76252-800 76250-096 76250-932 76249-248
Drill Monster® Jobbers Length Twist Drills, Cutting Tool Supply
Tool Sets
These general-purpose jobber length twist drills are designed in a manner to assure true, accurate and smooth holes.
  • 118°- drill point style
  • 25° helix - RHS/RHC (Right Hand Cut/Right Hand Spiral)
  • Diameter Tolerance: +0.0000/−0.0005"
  • Available in fractional, letter and wire gage size
These drills are made of solid carbide (C-2 grade premium sub-micron carbide with 10% cobalt), which provides higher wear resistance.
Ideal for use on non-ferrous alloys, cast iron, plastics, aluminum, and highly abrasive materials.
Ordering information: Available as either with or without coating (TiN or TiAlN coated).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty