Would you like to log in again?
Chemical Resistant Red Sable Brushes, Gordon Brush
Supplier: Gordon Brush
0907-06000 0907-00300 0907-00000 0907-02000 0907-04000
76243-092EA 27.33 USD InStock
76243-092 76243-090 76244-170 76241-972 76241-970
Chemical Resistant Red Sable Brushes, Gordon Brush
Brushes
These pure red sable brushes are ideal for applying conformal coatings.
  • Unvarnished hardwood handle, so chemicals will not cause paint to peel or flake
  • One of the world's most coveted artist brushes
  • Anti-static
The brushes are made to endure harsh chemicals, including toluene, without deterioration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty