Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AGAP2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-DN8
75931-000EA 518 USD InStock
75931-000
Anti-AGAP2 Rabbit Polyclonal Antibody
Antibodies
Phosphoinositide 3 kinase enhancer (PIKE) is a recently identified nuclear GTPase that interacts with nuclear phosphoinositide 3-kinase (PI3 kinase) to stimulate its lipid kinase activity. PIKE exists in multiple isoforms; a shorter C-terminal isoform (PIKE-A) has also been identified as centaurin gamma 1. The longest isoform (PIKE-L) has been shown to bind to the adaptor protein Homer and thereby link to metabotropic glutamate receptors, leading to activation of PI3 kinase activity and prevention of neuronal apoptosis. Overexpression of PIKE-A enhances Akt activity and promotes cancer cell invasion, whereas decreased expression of PIKE-A via dominant negative expression of PIKE-A or PIKE-A knockdown inhibits these processes. In many human cancers, expression of PIKE-A is enhanced, leading to increased Akt activity and preventing apoptosis.

Anti-PIKE Antibody has been tested for use in ELISA, Western Blotting, and Immunohistochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 125 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: AGAP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty