Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DDX58 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

600-401-H15
75931-618EA 518 USD InStock
75931-618
Anti-DDX58 Rabbit Polyclonal Antibody
Antibodies
RIG-1 antibody detects the human RIG-1 protein. The innate immune system detects viral infection by recognizing various viral components and triggers antiviral responses. Like the toll-like receptor 3 (TLR3), the cytoplasmic helicase retinoic acid inducible gene protein 1 (RIG-1) recognizes double-stranded (ds) RNA, a molecular pattern associated with viral infection. Unlike TLR3 however, RIG-1 activates the kinases TBK1 and IKKε through the adaptor protein IPS-1. These kinases then phosphorylate the transcription factors IRF-3 and IRF-7 which are essential for the expression of type-I interferons. RIG-1 is required for the production of interferons in response to RNA viruses including paramyxoviruses, influenza virus, and Japanese encephalitis virus. Anti-RIG-1 antibodies are ideal for investigators involved in infectious disease research.

Anti-RIG-1 Antibody is suitable for use in E,WB, and IHC. Expect a band approximately ~106.6 kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: DDX58
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty