Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GPR3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BM9
75928-180EA 518 USD InStock
75928-180
Anti-GPR3 Rabbit Polyclonal Antibody
Antibodies
GPR3, also known as ACCA, is a G-protein coupled receptor that constitutively activates adenylate cyclase and is highly expressed in the central nervous system. Overexpression of GPR3 stimulates the production of amyloid-beta peptide (Abeta), the deposition of which is one of the pathological hallmarks of Alzheimer 's disease (AD), while the ablation of GPR3 prevented the accumulation of Abeta in vitro and in an AD mouse model. This is of particular interest because of the proximity of a reported candidate Alzheimer 's disease (AD) locus, suggesting that GPR3 may be a potential therapeutic target for the treatment of AD. GPR3 has also been shown to block the proliferation of cerebellar granule cell precursors (GCP) during postnatal development by inhibiting the Shh-induced proliferation of GCP, indicating that GPR3 activation may represent one of the signals that triggers the postnatal cell cycle exit and terminal differentiation of GPCs.

Anti-GPR3 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 35 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: GPR3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty