Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IFIT1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BT2
75934-628EA 518 USD InStock
75934-628
Anti-IFIT1 Rabbit Polyclonal Antibody
Antibodies
The interferon-induced protein with tetratricopeptide repeats 1 (IFIT1) protein is a member of a family of tetratricopeptide repeat-containing proteins whose transcription is upregulated by interferons, virus infection, and molecular patterns such as dsRNA or lipopolysaccharides (1,2). These proteins have been suggested to induce anti-viral cellular activities in response to infection (2). Together with the interferon-induced transmembrane protein 1 (IFITM1), IFIT1 inhibits the replication of the Hepatitis C virus, suggesting that it may be useful in the development of therapeutic treatments (3).

Anti-IFIT1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 55 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: IFIT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty