Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL-10 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
109-401-312-0100
76234-816EA 530 USD InStock
76234-816
Anti-IL-10 Rabbit Polyclonal Antibody
Antibodies
Anti IL-10 Antibody recognizes IL-10 (IL-10 or IL10), also known as human cytokine synthesis inhibitory factor (CSIF), that is an anti-inflammatory cytokine. In humans IL-10 is encoded by the IL10 gene. IL-10 is produced primarily by monocytes and to a lesser extent by lymphocytes. This cytokine has pleiotropic effects in immunoregulation and inflammation. It down-regulates the expression of Th1 cytokines, MHC class II antigens, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. IL-10 can block NF-kB activity, and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice suggested the function of this cytokine as an essential immunoregulator in the intestinal tract and indeed patients with Crohn's disease react favorably towards treatment with bacteria producing recombinant interleukin 10, showing the importance of interleukin 10 for counteracting excessive immunity in the human body. Anti-IL-10 cytokine antibody is ideal for investigators involved Immunology and Signal Transduction research.

This antiserum has been tested for use in neutralizations, ELISA, immunohistochemistry, radioimmunoassay, immunoprecipitation, immunohistochemistry and immunoblotting.  The antibody recognizes huIL-10.  Reactivity in other immunoassays is unknown.

Type: Primary
Antigen: IL-10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty