Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Irak2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BZ3
75929-224EA 518 USD InStock
75929-224
Anti-Irak2 Rabbit Polyclonal Antibody
Antibodies
The pro-inflammatory cytokine IL-1 induces cellular response through two subunits of its receptor, IL-1 receptor I (IL-1RI) and IL-1 receptor accessory protein (IL-1RAcP). IL-1 receptor-associated kinase (IRAK) mediates activation of NF-kB, which is a pivotal transcription factor mediating inflammatory and immune response. A novel member in the IRAK/Pelle family has been identified and designated IRAK2. Both IRAK and IRAK2 recruit to the subunits of the IL-1R complex after IL-1 binding and lead to NF-kB activation. IRAKs also associate with Toll like receptor (TLR) and the dominant negative mutants of IRAKs inhibit LPS-induced NF-kB activation. Members in IRAK/Pelle family play a central role in IL-1R and TLR mediated inflammatory response. Unlike human IRAK2, murine IRAK2 exists as four alternately spliced isoforms (IRAK2a-d), with two isoforms (IRAK2c and d) acting in an inhibitory fashion. IRAK2 is expressed in a variety of tissues.

Anti-IRAK2 Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 69 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: Irak2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty