Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IVL Mouse Monoclonal Antibody [Clone: IL-9]

Supplier: Rockland Immunochemicals
200-301-P39
75929-188EA 528 USD InStock
75929-188
Anti-IVL Mouse Monoclonal Antibody [Clone: IL-9]
Antibodies
Involucrin is a keratinocyte protein that first appears in the cell cytosol, but ultimately becomes cross-linked to membrane proteins by transglutaminase. The gene consists of 585 amino acids, 390 of which form a central decapeptide repeat, rich in glutamine and glutamic acid. The involucrin gene, encoding a protein of the terminally differentiated keratinocyte, is polymorphic in the human. Involucrin has previously been mapped to chromosome 1q21. The epidermal protein involucrin has been remodeled in the higher primates. And the existence of two populations of East Asian involucrin alleles gives support for the existence of a Eurasian stem lineage from which Caucasians and a part of the Asian population originated. This antibody is suitable for researchers interested in skin cell research.

Anti-Involucrin is useful for Immunohistochemistry -P, -F, Immunocytochemistry, and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~68.5 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: IVL
Clonality: Monoclonal
Clone: IL-9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty