Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KDM4A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X45
75929-322EA 487 USD InStock
75929-322
Anti-KDM4A Rabbit Polyclonal Antibody
Antibodies
JMJD2a, belongs to the JMJD2 family of histone demethylases which play an important role in the establishment of the histone code. JMJD2a specifically demethylates the trimethylated K9 and K36 of histone H3, thereby converting these lysines to the dimethylated form. It has no activity towards H3K4, H3K27 and H4K20, or to the mono- and dimethylated H3K9 and H3K36. JMJD2a plays a role in the transcriptional repression of ASCL2 and E2F-responsive promoters via the recruitment of histone deacetylases and NCOR1, respectively. Anti-JMJD2a Antibody is ideal for research in Chromatin Remodeling, Gene Expression and Epigenetics.

Anti-JMJD2a Antibody is suitable for ELISA and Western Blots. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 121 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: KDM4A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty