Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KLF4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GU2
76262-954EA 518 USD InStock
76262-954
Anti-KLF4 Rabbit Polyclonal Antibody
Antibodies
KLF4 belong to the Kruppel like factors/Zinc fingers C2H2-type family. This gene encodes a protein that belongs to the Kruppel family of transcription factors. KLF4 can act both as activator and as repressor. It can bind the 5-CACCC-3 core sequence, the promoter region of its own gene, and can activate its own transcription. KLF4 regulates the expression of key transcription factors during embryonic development, playing an important role in maintaining embryonic stem cells, and in preventing their differentiation. This protein is required for normal development of the barrier function of skin postnatal, maintenance of the ocular surface, function in skeletal and kidney development, and thought to control the G1-to-S transition of the cell cycle following DNA damage by mediating the tumor suppressor gene p53. KLF4 may be associated with diseases such as Venous Hemangioma, Skin Squamous Cell Carcinoma, and pediatric lymphoma. Anti-KLF4 Antibody is useful for researchers interested in Peptide hormone metabolism, Stem Cell Differentiation Pathway Research, Wnt/Notch Pathway Research, and Cancer Research.

Anti-KLF4 Antibody has been tested in Western Blot, ELISA, and Immunofluorescence. Expect a band at ~54.7 kDa in western blot using appropriate lysates. Positive control whole cell lysates used K562 and PC3 @ 1µg/mL for WB; and PC3 with PFA at 10µg/mL in IF. Although not tested, this antibody is likely functional in immunohistochemistry, flow cytometry, and immunoprecipitation.

Type: Primary
Antigen: KLF4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty