Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MUC5B Mouse Monoclonal Antibody [Clone: MG-31]

Supplier: Rockland Immunochemicals
200-301-P41
75930-158EA 528 USD InStock
75930-158
Anti-MUC5B Mouse Monoclonal Antibody [Clone: MG-31]
Antibodies
MUC1 is a large cell surface mucin glycoprotein expressed by most glandular and ductal epithelial cells and some hematopoietic cell lineages. It is expressed on most secretory epithelium, including mammary gland and some hematopoietic cells. It is expressed abundantly in lactating mammary glands and overexpressed abundantly in >90% breast carcinomas and metastases. Transgenic MUC1 has been shown to associate with all four cebB receptors and localize with erbB1 (EGFR) in lactating glands. The MUC1 gene contains seven exons and produces several different alternatively spliced variants. The major expressed form of MUC1 uses all seven exons and is a type 1 transmembrane protein with a large extracellular tandem repeat domain. The tandem repeat domain is highly O glycosylated and alterations in glycosylation have been shown in epithelial cancer cells. This antibody is suitable for researchers interested in neuroscience research, developmental signaling, and cell adheshion.

Anti-Mucin Gastric is useful for Immuohistochemistry -P, Immunocytochemistry, and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~596 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: MUC5B
Clonality: Monoclonal
Clone: MG-31
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty