Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NOSTRIN Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DD6
75934-954EA 518 USD InStock
75934-954
Anti-NOSTRIN Rabbit Polyclonal Antibody
Antibodies
NOSTRIN. also known as endothelial nitric oxide synthase traffic inducer, is a member of the Pombe Cdc15 homology (PCH) family of proteins which may function as the modulator of the subcellular localisation and activity of endothelial nitric oxide synthase (eNOS) (1,2). It decreases NOS3 activity by inducing its translocation away from the plasma membrane (3). NOSTRIN typically exists as a trimer and is expressed in the vascular endothelial cells of highly vascularized tissues such as placenta, lung, kidney and heart. NOSTRIN is necessary for proper vascular development in zebrafish and postnatal retinal angiogenesis in mice (4,5).

Anti-NOSTRIN Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 58 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NOSTRIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty