Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-OASL Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DF6
75934-028EA 518 USD InStock
75934-028
Anti-OASL Rabbit Polyclonal Antibody
Antibodies
OASL (2'-5'-oligoadenylate synthetase-like), also known as p59OASL or TRIP14 (thyroid receptor-interacting protein 14), is a 514 amino acid protein that exists as two alternatively spliced isoforms, designated p56 and p30, and contains two ubiquitin-like domains (1). It is widely expressed in a variety of tissues and interacts with the ligand binding domain of the thyroid receptor (TR) and is able to bind double-stranded RNA and DNA, possibly playing a role in RNA degradation and the overall inhibition of protein synthesis (2-3). Methyl CpG-binding protein 1 (MBD1), which functions as a transcriptional repressor, was identified as a strong p59 OASL interactor (4).

Anti-OASL Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 59 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: OASL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty