Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-OTUD5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DH7
75930-656EA 518 USD InStock
75930-656
Anti-OTUD5 Rabbit Polyclonal Antibody
Antibodies
OTUD5 is a member of the OTU (ovarian tumor) domain-containing cysteine protease superfamily. The OTU domain confers deubiquitinase activity and OTUD5 has been shown to suppress the type I interferon (IFN-I)-dependent innate immune response by cleaving the polyubiquitin chain from TRAF3, an essential type I interferon adaptor protein. Cleavage results in disassociation of TRAF3 from a downstream signaling complex containing TBK1 and the disruption of the IFN-I signaling cascade, indicating that OTUD5 acts as a negative regulator of innate immune responses. It has been suggested that by suppressing IFN-I production, OTUD5 may function to inhibit the emergence of certain autoimmune disorders such as systemic lupus erythematosus. Multiple isoforms of OTUD5 are known to exist.

Anti-OTUD5 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 61 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: OTUD5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty