Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PHC3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DM0
75930-864EA 518.29 USD InStock
75930-864
Anti-PHC3 Rabbit Polyclonal Antibody
Antibodies
Polycomb group (PcG) proteins assemble into multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes throughout development (1). PHC3 (Polyhomeotic-like protein 3), also known as Early development regulatory protein 3, is a 983 amino acid nuclear protein that is a component of the PcG multiprotein PRC1 complex (2). PRC1 complex acts via chromatin remodeling and modification of histones. PHC3 contains 1 FCS-type zinc finger and SAM (sterile alpha motif) domain (2-3). PHC 3 has been regarded as a candidate tumor suppressor of osteosarcoma (4).

Anti-PHC3 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 106 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PHC3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty