Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PKM Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GV5S
76262-732EA 148 USD InStock
76262-732
Anti-PKM Rabbit Polyclonal Antibody
Antibodies
Anti-PKM2 Antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Pyruvate kinase has been shown to be expressed in tumor, embryonic and normal proliferating cells in varying levels. Studies show that when PKM2 is secreted it can induce EGFR phosphorylation which stimulates the EGFR downstream pathway. Previously PKM has been shown to be critical for aerobic glycolysis. Four isoforms of PKM play alternative roles within the system and post-translational modifications along these isoforms induce multiple pathways making this Anti-PKM2 antibody ideal for investigators involved in cancer research and post-translational modifications.

This affinity purified antibody has been tested for use in western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band 56-58 kDa in size corresponding to PKM2 by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: PKM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty