Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PUUVsMgp1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DV5
75935-060EA 518 USD InStock
75935-060
Anti-PUUVsMgp1 Rabbit Polyclonal Antibody
Antibodies
Puumala virus (PUUV) is a rodent-borne hantavirus of the family Bunyaviridae, an enveloped, negative-sense RNA viruses with a tripartite genome that can cause hantavirus pulmonary syndrome (HPS) and is highly homologous to the protype hantavirus Hantaan virus (1). Like other hantaviruses, the PUUV glycoprotein is synthesized as a precursor that is posttranslationally processed into two glycoproteins G1 (Gn) and G2 (Gc). These glycoproteins interact with the PUUV nucleocapsid (NP) protein through their cytoplasmic tail, and this association has been suggested to be crucial to the binding of the ribonucleoprotein of the PUUV and the assembly of the virus particle (2).

Anti-Puumala Virus Glycoprotein Antibody has been tested for use in ELISA. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 126 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PUUVsMgp1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty