Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RHOG Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DY9
75931-596EA 518 USD InStock
75931-596
Anti-RHOG Rabbit Polyclonal Antibody
Antibodies
The ras homolog family member G (RHOG), also known as rho-related GTP-binding protein, is a member of the Rho family of small GTPases, which cycle between inactive GDP-bound and active GTP-bound states and function as molecular switches in signal transduction cascades (1). RHOG controls a pathway that requires the microtubule network and activates Rac1 and Cdc42Hs independently of their growth factor signaling pathway (2). RHOG is also involved in the transcriptional regulation of interferon-gamma and nuclear factor of activated T cells (NFAT) and the regulation of the actin skeleton in lymphocytes (3).

Anti-RHOG Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 21 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: RHOG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty