Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SAMHD1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-ED7
75931-718EA 518 USD InStock
75931-718
Anti-SAMHD1 Rabbit Polyclonal Antibody
Antibodies
The SAM domain and HD domain 1 (SAMHD1) protein is upregulated in response to viral infection and is thought to play a role in innate immunity (1). SAMHD1 blocks the infection of HIV-1 and SIVdeltaVpx before reverse transcription in macrophages and dendritic cells (2), and this restriction is regulated by phosphorylation of SAMHD1 (3). Mutations in this gene have been associated with Aicardi-Goutieres syndrome (1).

Anti-SAMHD1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 72 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SAMHD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty