Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SDHAF2 Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-901-EF4
75931-764EA 518 USD InStock
75931-764
Anti-SDHAF2 Chicken Polyclonal Antibody
Antibodies
SDHAF2 (Succinate dehydrogenase complex assembly factor 2) encodes a mitochondrial protein needed for FAD cofactor attach to the SDH enzyme which plays a critical role in mitochondria. SDHAF2 is a tumor suppressor, but recent evidence suggests that somatic mutations of the SDHAF2 are unlikely to contribute to parathyroid tumor development in sporadic primary hyperparathyroidism.

Anti-SDHAF2 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 20 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SDHAF2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty