Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SMURF2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-ER7
75932-056EA 518 USD InStock
75932-056
Anti-SMURF2 Rabbit Polyclonal Antibody
Antibodies
SMURF2 (SMAD ubiquitin regulatory factor 2) is a negative regulator of TGF-beta signaling (1). SMURF1 and SMURF2 are members of HECT domain E3 ubiquitin ligase which are involved in the enzymatic reactions of the Ub conjugating pathway (1,2). SMURF2 is widely expressed and was initially identified as an inhibitor of TGF-beta/BMP signaling by targeting R-Smads and TGF type I receptor for ubiquitination and degradation (3). Studies have shown that SMURF2 functions as a tumor suppressor by maintaining genomic stability through targeting RNF20 (3). SMURF2 associates constitutively with SMAD7 (4).

Anti-SMURF2 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 87 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SMURF2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty