Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STAT5A Mouse Monoclonal Antibody [Clone: 5F6.F1]

Supplier: Rockland Immunochemicals
200-301-B18
75932-208EA 2704 USD InStock
75932-208
Anti-STAT5A Mouse Monoclonal Antibody [Clone: 5F6.F1]
Antibodies
Signal transducer and activator of transcription 5 (Stat5) belongs to a family of cytoplasmic transcription factors that can be activated (phosphorylated) by a cell surface receptor. Phosphorylation at Tyr694 is obligatory for Stat5 activation. Stat5 has two isoforms, Stat5 alpha and Stat5 beta. Aberrant Stat5 activation has been implicated in the pathogenesis of chronic myelogenous leukemia, prostate and breast cancer and tumor metastasis. Stat5 is localized in the cytoplasm and upon phosphorylation at Y694 is translocated to the nucleus. Ideal for Cancer, Chromatin & Nuclear Signaling and Signal Transduction research.

This monoclonal antibody is suitable for ELISA, immunohistochemistry and western blotting.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 91 kDa in size corresponding to phosphorylated Stat5a protein by western blotting in the appropriate cell lysate or extract.  This phospho-specific monoclonal antibody reacts with mouse Stat5a pY694 and shows minimal reactivity by ELISA against the non-phosphorylated form of the immunizing peptide.

Type: Primary
Antigen: STAT5A
Clonality: Monoclonal
Clone: 5F6.F1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty