Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRIM24 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FN9
75933-104EA 518 USD InStock
75933-104
Anti-TRIM24 Rabbit Polyclonal Antibody
Antibodies
TRIM24 (TIF1), a member of the tripartite motif (TRIM) family, plays a role in the regulation of cell proliferation and apoptosis. TRIM24 localizes to nuclear bodies and is thought to associate with chromatin and heterochromatin-associated factors. The TRIM motif includes three zinc-binding domains (RING, B-box type 1 and B-box type 2) and a coiled-coil region. TIF1 mediates transcriptional events by interactions with the AF2 region of several nuclear receptors, such as the estrogen, retinoic acid and vitamin D3 receptors. Defects in TRIM24 are a cause of thyroid papillary carcinoma (TPC).

Anti-TRIM24 Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 117 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TRIM24
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty