Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-UNG Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FV1
75933-448EA 518 USD InStock
75933-448
Anti-UNG Rabbit Polyclonal Antibody
Antibodies
The human uracil-DNA glycosylase (UNG) gene encodes both mitochondrial (UNG1) and nuclear (UNG2) forms through differentially regulated promotes and alternative splicing. While UNG2 is the major enzyme in the base excision repair pathway that removes uracil residues from nuclear DNA that arise through either misincorporation during replication or cytosine deamination, inhibition of UNG1 by uracil glycosylase inhibitor did not lead to increased levels of spontaneous or induced mitochondrial DNA mutations. However, decreased levels of UNG activity and increased oxidative damage to mitochondrial DNA were seen in older mice, suggesting that mitochondrial DNA repair mechanisms may be involved in various neurodegenerative disorders in an age-dependent manner. This UNG1 antibody will not cross-react with UNG2.

Anti-UNG1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 34 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: UNG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty