Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-USP25 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FV6
75933-458EA 518 USD InStock
75933-458
Anti-USP25 Rabbit Polyclonal Antibody
Antibodies
USP25 (ubiquitin specific peptidase 25), also known as USP21, belongs to the peptidase C19 family and is a highly conserved 76-amino acid protein involved in regulation of intracellular protein breakdown, cell cycle regulation, and stress response (1). It contains one UBA-like domain and two UIM (ubiquitin-interacting motif) repeats. Due to alternative splicing events, USP25 is expressed as two short, ubiquitously expressed isoforms and one long, muscle-specific isoform (2). The long isoform of USP25 (USP25m) is upregulated in myogenesis and is implicated in regulation of muscular differentiation and function. USP25 is a deubiquitinating enzyme (DUB) that negatively regulates IL-17-triggered signaling (3,4).

Anti-USP25 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 122 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: USP25
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty