Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-WNVgp1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FY6
75933-622EA 518 USD InStock
75933-622
Anti-WNVgp1 Rabbit Polyclonal Antibody
Antibodies
West Nile Virus (WNV) is a member of the Flaviviridae, a plus-stranded virus family that includes St. Louis encephalitis virus, yellow fever virus, and Dengue virus. WNV was initially isolated in 1937 in the West Nile region of Uganda and has become prevalent in Africa, Asia, and Europe. It has rapidly spread across the United States with cases being observed in every continental state. Virus particles consist of a dense core made up of the core/capsid protein encapsulating the RNA genome surrounded by a membrane envelope embedded with envelope and matrix proteins. While the viral core protein is thought to contribute to the WNV-associated inflammation via apoptosis induced though the caspase-9 pathway, the highly glycosylated envelope protein plays a major role for WNV entry into target cells as this entry can be inhibited by using a recombinant domain III from the envelope glycoprotein. The WNV receptor has recently been identified as alpha v beta 3 integrin.

Anti-West Nile Virus Envelope Antibody has been tested for use in ELISA. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 380 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: WNVgp1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty