Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-XBP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-H33
75935-492EA 518 USD InStock
75935-492
Anti-XBP1 Rabbit Polyclonal Antibody
Antibodies
XBP-1 antibody detects human XBP-1. X box binding protein 1 (XBP-1) is a key protein in the mammalian unfolded protein response (UPR) that protects the cell against the stress of malfolded proteins in the endoplasmic reticulum (ER). Upon sensing unfolded proteins, an ER transmembrane endonuclease and kinase termed IRE1p is activated and excises an intron from XBP-1 mRNA. The spliced XBP-1 mRNA results in a 371 amino acid protein (XBP-1s) which is then translocated to the nucleus where it binds to the regulatory elements of downstream genes. Together with other UPR transcription factors such as ATF6, XBP-1 stimulates the production of ER stress proteins including the ER resident protein chaperones glucose regulated protein (GRP) 78 and GRP94. Anti-XP1 is ideal for investigators involved in enzyme, cell cycle proteins and cancer research.

Anti-XBP-1 Antibody is suitable for use in ELISA, Western Blot, Immunofluorescence, and Immunohistochemistry. Expect a band approximately ~28.6kDa using positive control HepG2 whole cell lysates, which also can be used for ICC and IF, or other specific lysates. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: XBP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty