Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ZMYM4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GF9
75933-760EA 518 USD InStock
75933-760
Anti-ZMYM4 Rabbit Polyclonal Antibody
Antibodies
Zinc-finger proteins contain DNA-binding domains characterized by the unique role of zinc and have a wide variety of functions such as transcriptional activation or repression. The protein folding and the DNA binding ability are governed by the coordination of a zinc ion. It has been found to be overexpressed in human lung adenocarcinomas and squamous cell carcinomas, and the overexpression of a fragment of the 3'UTR of the ZMYM4 mRNA termed Cell Death Inhibiting RNA (CDIR) protects HeLa cells from IFN-gamma-induced apoptosis, suggesting that ZMYM4 may play a role in tumorigenesis.

Anti-ZMYM4 Antibody has been tested for use in ELISA, and Immunohistochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 173 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: ZMYM4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty