Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG1 Goat Mouse Polyclonal Antibody (BAC)

Supplier: Rockland Immunochemicals
610-106-040
75933-930EA 252 USD InStock
75933-930
Anti-IgG1 Goat Mouse Polyclonal Antibody (BAC)
Antibodies
Anti-Mouse IgG1 Biotin Antibody generated in goat detects reactivity to Mouse IgG1 (Gamma 1 chain). Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. IgG1 chain constitutes 66% of the IgG subclass and has a high affinity for binding to the Fc receptor of phagocytic cells. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition.

Mouse IgG1 Biotin Conjugated Secondary Antibodies are ideal for western blotting, Immunohistochemistry and ELISA as well as other antibody detection methods.

Type: Secondary
Antigen: IgG1
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty