Would you like to log in again?
Transverse End Cutters, Swanstrom Tools USA
Supplier: Swanstrom Tools USA
S233E-TP
76241-346EA 179.34 USD InStock
76241-346
Transverse End Cutters, Swanstrom Tools USA
Cutters
These ergonomic style transverse end cutters are ESD safe (static dissipative) which protects circuit components.
  • Does not insulate
  • Do not use on live circuits
These superior quality tools are forged from high chromium, high carbon alloy steel.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty