Would you like to log in again?
Round Nose Pliers, Swanstrom Tools USA
Supplier: Swanstrom Tools USA
S236E-TP
76243-552EA 130.71 USD InStock
76243-552
Round Nose Pliers, Swanstrom Tools USA
Pliers
These ergonomic style round nose pliers are ESD safe (static dissipative) which protects circuit components.
  • Does not insulate
  • Do not use on live circuits
These superior quality tools are forged from high chromium, high carbon alloy steel.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty