Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium carbonate heavy 40.0 - 43.5%, powder USP, FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Magnesium carbonate hydroxideMagnesium carbonate basic, heavy

Management of Change (MOC) category = R

MACR5946-32 MACR5946-32
MK5946-32EA 930.69 USD InStock
CAMK5946-32 MK5946-32
Magnesium carbonate heavy 40.0 - 43.5%, powder USP, FCC, Macron Fine Chemicals™
Magnesium carbonate heavy


Formula: CH₂Mg₂O₅
MW: 142,63 g/mol
Density: 3.03 g/cm³ (16 °C)
MDL Number: MFCD00011107
CAS Number: 39409-82-0
EINECS: 208-915-9
Order Now
Specification Test Results
GMP Manufactured Product
Meets F.C.C. Requirements
Meets U.S.P Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
Assay (MgO) 40.0 - 43.5 %
Appearance (bulky, white powder) Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
Microbiological Passes Test
Soluble Salts ≤ 1.0 %
Acid-Insoluble Substances ≤ 0.05 %
Solubility in Dilute Sulfuric Acid Passes Test
Limit of Calcium (Ca) ≤ 0.45 %
Iron (Fe) ≤ 0.02 %
Calcium Oxide ≤ 0.6 %
Lead (Pb) ≤ 2 mg/kg
Arsenic (As) ≤ 3 ppm
Bulk Density (g/cc) ≥ 0.34
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty