Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
HFC35 Series Couplings, Colder Products Company
Supplier: CPC (Colder Products Company)
HFCD22635 HFC24835 HFCD23835 HFC16835 HFCD22635 HFCD10835 HFCD17835 HFCD23635 HFCD221035 HFCD16835 HFC10835 HFCD171035 HFCD17635 HFCD24835 HFC221035 HFCD22835
76254-092EA 20.16 USD InStock
76254-092 76254-118 76240-870 76240-872 76254-116 76240-864 76254-132 76254-120 76254-098 76254-134 76240-866 76240-868 76254-094 76277-636 76254-130 76254-096
HFC35 Series Couplings, Colder Products Company
Fittings and Connectors
HFC35 series polysulfone couplings are able to withstand demanding applications and feature physical strength, chemical resistance and autoclavability.
  • Operating temperature: −40 to 138 °C
  • Easy to grip, simple to operate
  • Physically strong and UV resistant
  • Valve spring is made of 316 stainless steel
An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty