Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 50% in aqueous solution, AR®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

Management of Change (MOC) category = NI

MACR7706-68 MACR7706-68
MK7706-68EA 10232.84 USD InStock
CAMK7706-68 MK7706-68
Sodium hydroxide 50% in aqueous solution, AR®, Macron Fine Chemicals™
Sodium hydroxide
Caution: Protect from air to avoid absorption of carbon dioxide.


Formula: NaOH
MW: 40 g/mol
Density: 1.53 g/cm³
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Protect from air to avoid absorption of carbon dioxide.
Assay (NaOH) 49.5 - 52.0 %
Identification Passes Test
Ammonium Hydroxide Precipitate ≤0.010 %
Chloride (Cl) ≤0.0025 %
Heavy Metals (as Ag) ≤0.0015 %
Iron (Fe) ≤0.0005 %
Nitrogen Compounds (as N) ≤0.0005 %
Potassium (K) ≤0.025 %
Phosphate (PO₄) ≤0.0005 %
Sulfate (SO₄) ≤0.0025 %
Sodium Carbonate (Na₂CO₃) ≤0.1 %
Appearance (clear, colorless liquid)

 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty