Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Safety Wire Twisters, Jenson Tools
Supplier: Jenson Tools
6WT 1W393
76243-576EA 139.12 USD InStock
76243-576 76243-968
Safety Wire Twisters, Jenson Tools
Pliers
Safety wire twisters are used to grip, twist, and cut safety wire.
  • Made of high carbon alloy steel
  • Black oxide finish
Tapered nose for tough to reach areas.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty