Would you like to log in again?
Puritan® Standard Polyester Swab, Hardwood Products Company
Supplier: Hardwood Products Company
25-8061PD
76237-982CS 263.9 USD InStock
76237-982
Puritan® Standard Polyester Swab, Hardwood Products Company
Swabs Swabs for Clinical Use
Regular polyester tipped applicator with semi-flexible plastic handle.
Ideal for specimen collection, cell collection for DNA testing, collecting specimens from throats, gynecological screening, industrial surface testing.
Delivery information: Individually wrapped.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty