Would you like to log in again?
PurSwab® Lint Free Double-Ended Cotton Tipped Swab, Hardwood Products Company
Supplier: Hardwood Products Company
893-PCDBL 870-PCDBL 891-PCDBL
76241-170CS 324.94 USD InStock
76241-170 76244-820 76239-994
PurSwab® Lint Free Double-Ended Cotton Tipped Swab, Hardwood Products Company
Swabs Swabs for Clinical Use
Double sharp pointed cotton tipped applicator with paper handle.
  • Highly absorbent and does not swell in solvents
  • Lint and residue free cotton swab
  • Sharp, double pointed tip with fine paper handle
  • Anti-static packaging contains no silicon
Fine pointed, double tip is ideal for cleaning sensitive components. Ideal for critical envrionments, low in NVR.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty