Would you like to log in again?
Hospital® Standard Cotton Swab with Wooden Handle, Hardwood Products Company
Supplier: Hardwood Products Company
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
806-WCHOSPITAL
76240-702CS 264.66 USD InStock
76240-702
Hospital® Standard Cotton Swab with Wooden Handle, Hardwood Products Company
Swabs Swabs for Clinical Use
Economy grade regular cotton tipped applicator with wood handle.
  • Regular cotton head size on rigid wood handle
All purpose swab, ideal for general cleaning in medical or industrial settings, cleaning instrumentation and surfaces, applying solder paste and removing excess solder/fluxes from circuit boards
removing excess from injection molding, removing oils; paints; and dirt from machine parts
used in patient care for cleaning wounds or applying topical dressings, cosmetic applications.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty