Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
PurSwab® Small Soft Foam Swab with Polypropylene Handle, Hardwood Products Company
Supplier: Hardwood Products Company
1105
76253-562CS 1781.97 USD InStock
76253-562
PurSwab® Small Soft Foam Swab with Polypropylene Handle, Hardwood Products Company
Swabs Swabs for Clinical Use
Soft non-abrasive foam tipped applicator with plastic handle.
  • Lint and residue free tip
  • Thermally bonded tip reduces contaminates
  • Can be used with IPA or Deionized water or dry
  • Thickwalled sturdy handle for ease of handling
General use swab, ideal for use in critical environments. Used for applying adhesives and solvents. Soft non-abrasive tip ideal for use on electronics.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty