Would you like to log in again?
Excellence XSR Legal for Trade (NTEP) Precision Balances, METTLER TOLEDO®
Supplier: Mettler-Toledo
30317518 30316521 30317504 30317446 30317552 30317465 30316471 30317457 30317415 30317510
76098-050EA 5560 USD InStock
76098-050 76098-040 76098-144 76098-046 76098-146 76098-158 76098-052 76098-042 76098-054 76098-044
Excellence XSR Legal for Trade (NTEP) Precision Balances, METTLER TOLEDO®
Balances and Scales
XSR balances are easy and comfortable to use, protecting your health during your daily weighing tasks. The extraordinary stability provided by the new Monobloc™ weighing cell means you get your results fast even under challenging production environments.
  • 4.3" color capacitive touchscreen terminal, glove compatible
  • Integrated results notepad
  • Multiple interfaces for easy connectivity and data export
  • Easy connection of peripheral devices: printers, PC, barcode readers, footswitches and sensors
  • MinWeigh warning function
  • Password protected configuration
  • Integrated overload protection
  • Made of chemically resistant materials
  • Designed for easy cleaning
XSR precision balances offer a winning combination of reliable performance and truly ergonomic operation. Even under the harshest of weighing conditions, you can rely on these robust balances to help you meet your productivity targets.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty