Would you like to log in again?
Diamond Saw Lubricant
Supplier: COVINGTON ENGINEERING CORP
R80100
470310-428EA 38.79 USD InStock
470310-428
Diamond Saw Lubricant
Lubricants
This mineral based oil has no distasteful odor and features excellent cooling properties that extend the life of the blade and saw.
  • High flashpoint of 230° F ensures safe operation in all conditions
We recommend this oil for all saws over 10" for cutting hard stone.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty