Would you like to log in again?
NSH Series Couplings, Colder Products Company
Supplier: CPC (Colder Products Company)
NSHD24012 NSHD10012
76245-260PK 668.83 USD InStock
76245-260 76248-020
NSH Series Couplings, Colder Products Company
Fittings and Connectors
NSH Series couplings are general purpose versions of our pressure-balanced, non-spill, ChemQuik® CQG couplings. Their non-spill design virtually eliminates spills, minimizes downtime and enhances operator safety in a very low-cost package.
  • Operating temperature: 0 to 49 °C
  • Eliminates source of metallic contaminants
  • Helps prevent accidental misconnects
  • Ultimate protection from chemicals and fumes
Molded polypropylene, EPDM seals and a 100% springless and metal-free flow path provide broad chemical resistance and exceptionally high flow capacity, allowing instant disconnects (and reconnects), even under pressure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty