Would you like to log in again?
Wide-Jet Nozzle, Steinel America
Supplier: STEINEL AMERICA INC. TE
7021
76252-232EA 36.38 USD InStock
76252-232
Wide-Jet Nozzle, Steinel America
Heaters Hot Air Dryers
Wide jet nozzle spreads hot air over a wider area for drying, shaping plastic, heating large surfaces, and more.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty