Would you like to log in again?
Anti-Shock Shear Cutters, Excelta
Supplier: Excelta Corporation
EXA-530E-US-.0 530E-USS-020 EXA-530E-US-.0
76243-266EA 398.21 USD InStock
76243-266 76248-640 76253-040
Anti-Shock Shear Cutters, Excelta
Cutters
The 'Stand-Off' of these cutters keeps the cutting blade from cutting into the solder connections.
  • The grips are static dissipative
These cutters reduce shock on delicate components and solder connections.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty