Would you like to log in again?
Lazer Line™ Medium Cutter, Excelta
Supplier: Excelta Corporation
7147ES
76250-476EA 393.35 USD InStock
76250-476
Lazer Line™ Medium Cutter, Excelta
Cutters
This cutter features induction hardened cutting blades to RHC 63-65.
  • Grips are soft cushioned foam and are ESD safe
  • Dual leaf springs provide smooth action
The CNC machining allows the joint surfaces to be perfect to allow for long life.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty