Would you like to log in again?
High Precision Ergo-tek Slim Cutters, Large Tapered and Relieved Head, Full-Flush, ESD Safe, Ideal-tek
Supplier: Ideal-Tek
ES5552
76250-558EA 74.07 USD InStock
76250-558
High Precision Ergo-tek Slim Cutters, Large Tapered and Relieved Head, Full-Flush, ESD Safe, Ideal-tek
Cutters
As with all tools, it’s important to be application specific to ensure the best results. Ergo-tek slim cutters offer a complete solution to maximize production while ensuring worker safety. Ideal-tek cutters are specifically designed to meet customer requests. The overall handle design increases visibility and accessibility in confined areas and into tight spaces.
  • High-quality ball bearing steel
  • 63 Rockwell Hardness (HRC) for cutters
  • Superior no-scratch/anti-glare satin finish
  • Full-Flush cutting blades, for perfect cutting of soft wire
  • Two-component, user-friendly, ergonomic handles with soft, comfortable gripping surface
  • Lap joint with screw ensures a high level of strength and precision
  • Perfect symmetry
The Ergo-tek line represents the perfect combination of ultra-high cutting quality and durability, with stylish and ergonomic design. The Ergo-tek Slim range is designed especially for smaller hands, while providing the same ergonomic, ESD, and technical advantages expected from Ideal-tek. Upgraded features include a revised ergonomic handle to provide enhanced operator comfort and reduce repetitive motion injuries, overall lighter weight, and better accessibility and visibility in confined areas.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty