Would you like to log in again?
Kevlar® Cutter with Blue Grips, Swanstrom Tools
Supplier: Swanstrom Tools USA
Kevlar®
9180
76249-846EA 61.89 USD InStock
76249-846
Kevlar® Cutter with Blue Grips, Swanstrom Tools
Cutters
Designed and manufactured for cutting Kevlar® and other high strength fibers.
  • Abrasion resistance and stretch free nature
Handles are free of confining finger loops and return spring brings tool back to an open position after each cut. one of the blades is serrated and acts to prevent the fibers from sliding out of the front of the scissor when cutting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty